Assay Details
| Gene Symbol: Ephb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290753.1 | NP_001277682.1 | 7 - 8 | 1775 | 85 | |
| NM_010142.3 | NP_034272.1 | 7 - 8 | 1772 | 85 | ||
| GenBank mRNA | BC043088.1 | - | 7 - 8 | 1696 | 6418088 | 85 |
| BC062924.1 | - | 7 - 8 | 1720 | 85 | ||
| L25890.1 | - | 8 - 9 | 1614 | 85 | ||
| X76011.1 | - | 1 - 2 | 70 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephb2
Genomic Map