Assay Details
| Gene Symbol: Zmiz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005867.1 | NP_001005867.1 | 7 - 8 | 1128 | 50 | |
| NM_028601.3 | NP_082877.2 | 7 - 8 | 1224 | 50 | ||
| GenBank mRNA | BC024081.1 | - | 6 - 7 | 974 | 50 | |
| BC060652.1 | - | 7 - 8 | 1066 | 30354393 | 50 | |
| CA318375.1 | - | 2 - 3 | 303 | 50 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zmiz2
Genomic Map