Assay Details
| Gene Symbol: Camk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174053.1 | NP_001167524.1 | 11 - 12 | 1116 | 99 | |
| NM_001174054.1 | NP_001167525.1 | 12 - 13 | 1149 | 99 | ||
| NM_007595.5 | NP_031621.3 | 11 - 12 | 1116 | 99 | ||
| GenBank mRNA | AK046454.1 | - | 11 - 12 | 1117 | 99 | |
| AK158794.1 | - | 8 - 9 | 877 | 99 | ||
| AK163698.1 | - | 11 - 12 | 933 | 99 | ||
| BC024803.1 | - | 12 - 13 | 965 | 99 | ||
| BC080273.1 | - | 11 - 12 | 918 | 6851995 | 99 | |
| X63615.1 | - | 11 - 12 | 931 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camk2b
Genomic Map