Assay Details
| Gene Symbol: Ccser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164316.1 | NP_001157788.1 | 8 - 9 | 2400 | 72 | |
| NM_183310.2 | NP_899133.2 | 8 - 9 | 2452 | 72 | ||
| GenBank mRNA | AK048645.1 | - | 5 - 6 | 546 | 72 | |
| AK173231.1 | - | 7 - 8 | 1145 | 72 | ||
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as MTA transcript cluster(s) TC0600000725.mm.1
					            	This assay targets the same location as MTA transcript cluster(s) TC0600000725.mm.1| Set Membership: |  Ambion siRNA available  Amplicon length between 71 and 85  3' Most  Recommended Assay  Mus musculus  1700  Probe spans exons | 
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
					
						
							Agilent
					
						
							Affymetrix
					
						
							ABI
					
						
							Illumina
					
					
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
				
			| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24492099 | - | - | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24492101 | - | - | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24492100 | - | - | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24492097 | - | - | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953013 | AK173231.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953013 | NM_001164316.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953013 | NM_183310.2 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953008 | BC107398.2 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953008 | BC107399.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953008 | BC117738.2 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1953008 | AK031754.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998339 | AK048645.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998344 | AK173231.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998344 | NM_001164316.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1998344 | NM_183310.2 | mouse_transcriptome | 
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159129 | - | - | 
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148654 | - | - | 
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118531 | - | - | 
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105771 | - | - | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998344 | AK173231.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998344 | NM_001164316.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1998344 | NM_183310.2 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953013 | AK173231.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953013 | NM_001164316.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953013 | NM_183310.2 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953008 | BC107398.2 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953008 | BC107399.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953008 | BC117738.2 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1953008 | AK031754.1 | mouse_transcriptome | 
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109114 | - | - | 
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122679 | - | - | 
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131524 | - | - | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24492097 | - | - | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24492099 | - | - | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24492101 | - | - | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24492100 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492615 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495426 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495414 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495175 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492327 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05476200 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495402 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495417 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492616 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492686 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492626 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05296592 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492692 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492340 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492618 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492237 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05217327 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493738 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492303 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492325 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492240 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492360 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492196 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492332 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492319 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495169 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492701 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495415 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495413 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492837 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492624 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492691 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493747 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492528 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493737 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05473361 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492243 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492836 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495416 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492310 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492320 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492312 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05351596 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492307 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492334 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492828 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492513 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495430 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493746 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492838 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495400 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492534 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492820 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05504745 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492831 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05351578 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492198 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492339 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492688 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492827 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05802305 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05473360 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05248840 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495423 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05836331 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492321 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492305 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492530 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495425 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493744 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493735 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495409 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492203 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492510 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493748 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492694 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492318 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495173 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492357 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495178 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492818 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495180 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492516 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495422 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493745 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493742 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495167 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493733 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492697 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05727170 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492308 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492195 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492536 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492511 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05802306 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492356 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492522 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495171 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05099714 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492520 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495406 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495429 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05375962 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05217319 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492201 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492231 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492823 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492833 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492813 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492699 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492829 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492323 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495170 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05296586 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492622 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495404 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495411 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492619 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05146034 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493736 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492687 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495177 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492614 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492338 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492689 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492336 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492824 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492815 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492247 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495419 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492627 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492533 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05135247 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492313 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492245 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05351582 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492515 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492822 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492690 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492342 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492532 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492235 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24495183 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492238 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492242 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24493730 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24492246 | - | - | 
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05518368 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10538718 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10538719 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10538720 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10538716 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10538717 | - | - | 
| Affymetrix HT MG-430 PM Array Plate | 1445275_PM_at | - | - | 
| Affymetrix Mouse Gene 1.1 ST Array | 10538715 | - | - | 
| Mouse Genome 430 2.0 Array | 1437306_PM_at | - | - | 
| Mouse Genome 430 2.0 Array | 1445275_at | - | - | 
| Mouse Genome 430 2.0 Array | 1445275_PM_at | - | - | 
| Mouse Genome 430 2.0 Array | 1437306_at | - | - | 
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232035_PM_at | - | - | 
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232035_at | - | - | 
| Affymetrix Mouse Clariom S Array | PSR0600005588.mm.1 | TC0600000725.mm.1 | MTA TC | 
| Affymetrix Mouse Clariom S Array | PSR0600005588.mm.1 | NM_001164316.1.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005588.mm.1 | NM_001164316.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005588.mm.1 | NM_183310.2.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005588.mm.1 | NM_183310.2 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005593.mm.1 | TC0600000725.mm.1 | MTA TC | 
| Affymetrix Mouse Clariom S Array | PSR0600005593.mm.1 | NM_001164316.1.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005593.mm.1 | NM_001164316.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005593.mm.1 | NM_183310.2.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005593.mm.1 | NM_183310.2 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005591.mm.1 | TC0600000725.mm.1 | MTA TC | 
| Affymetrix Mouse Clariom S Array | PSR0600005591.mm.1 | NM_001164316.1.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005591.mm.1 | NM_001164316.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005591.mm.1 | NM_183310.2.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005591.mm.1 | NM_183310.2 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005592.mm.1 | TC0600000725.mm.1 | MTA TC | 
| Affymetrix Mouse Clariom S Array | PSR0600005592.mm.1 | NM_001164316.1.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005592.mm.1 | NM_001164316.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005592.mm.1 | NM_183310.2.1 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0600005592.mm.1 | NM_183310.2 | Refseq | 
| Mouse Expression Set 430 | 1445275_at | - | - | 
| Mouse Expression Set 430 | 1437306_at | - | - | 
| Mouse Expression Set 430 | 1445275_PM_at | - | - | 
| Mouse Expression Set 430 | 1437306_PM_at | - | - | 
| Murine Genome U74v2 Set | 165552_i_at | - | - | 
| Murine Genome U74v2 Set | 165552_PM_i_at | - | - | 
| Affymetrix HT Mouse Genome 430B Array | 1445275_at | - | - | 
| Affymetrix HT Mouse Genome 430B Array | 1437306_PM_at | - | - | 
| Affymetrix HT Mouse Genome 430B Array | 1445275_PM_at | - | - | 
| Affymetrix HT Mouse Genome 430B Array | 1437306_at | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 612646 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6200576 | - | - | 
| Illumina mouse-6 v1.0 expression beadchip | 6200576 | - | - | 
| Illumina mouseRef-8 v1.0 expression beadchip | 0006200576 | - | - | 
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2708756 | - | - | 
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2708756 | - | - | 
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2517462 | - | - | 
| mouse-6 v1.1 (Illumina) | scl0330336.2_298-S | - | - | 
| Illumina Sentrix mouse-6.v1 bead array | 6200576 | - | - | 
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0330336.2_298-S | - | - | 
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200576 | - | - | 
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2708756 | - | - | 
 Back To Top
 Back To Top
 Ccser1
Ccser1 Genomic Map
Genomic Map Back To Top
 Back To Top