Assay Details
| Gene Symbol: Cyp4x1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003947.1 | NP_001003947.1 | 4 - 5 | 489 | 60 | |
| GenBank mRNA | AJ786769.1 | - | 4 - 5 | 489 | 60 | |
| AK038526.1 | - | 465 | 60 | |||
| BC113125.1 | - | 4 - 5 | 488 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
oxidation-reduction process
molecular_function
monooxygenase activity
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
heme binding
metal ion binding
aromatase activity
biological_process
oxidation-reduction process
molecular_function
monooxygenase activity
iron ion binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
heme binding
metal ion binding
aromatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114580 | AK038526.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114580 | BC113125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114580 | NM_001003947.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P114580 | AJ786769.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93990 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646049 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 146809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80914 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198551 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563651 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289146 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832599 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128485 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 138928 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115489 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114580 | AK038526.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114580 | BC113125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114580 | NM_001003947.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P114580 | AJ786769.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223608 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23223629 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05366066 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115489 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00964581 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115489 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03833916 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03833919 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03505045 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03833925 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03833924 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03833917 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03505046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23223483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03815168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03815165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03422512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03888984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03422513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03215662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03888993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03833912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03422515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03943258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03505045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03815173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223597 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03833919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04041837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223663 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223628 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223586 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03988789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223656 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03815161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223600 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23223591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03815164 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10515179 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515177 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515170 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515178 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515173 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515180 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515171 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515169 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515176 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515174 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515175 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10515172 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441662_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10515168 | - | - |
| Mouse Genome 430 2.0 Array | 1441662_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1441662_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400026377.mm.1 | TC0400003311.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026377.mm.1 | NM_001003947.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026377.mm.1 | NM_001003947.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026376.mm.1 | TC0400003311.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400026376.mm.1 | NM_001003947.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400026376.mm.1 | NM_001003947.1 | Refseq |
| Mouse Expression Set 430 | 1441662_at | - | - |
| Mouse Expression Set 430 | 1441662_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441662_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441662_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 701894 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3163399 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3163399 | - | - |
| mouse-6 v1.1 (Illumina) | GI_51592064-A | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3163399 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3163399 | - | - |

Cyp4x1
Genomic Map