Assay Details
| Gene Symbol: Cyp4x1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003947.1 | NP_001003947.1 | 9 - 10 | 1197 | 63 | |
| GenBank mRNA | AJ786769.1 | - | 9 - 10 | 1197 | 63 | |
| AK038526.1 | - | 1173 | 63 | |||
| BC113125.1 | - | 9 - 10 | 1196 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp4x1
Genomic Map