Assay Details
Gene Symbol: Sec14l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146013.1 | NP_666125.1 | 6 - 7 | 585 | 85 | |
GenBank mRNA | AK028954.1 | - | 6 - 7 | 546 | 85 | |
BC026948.1 | - | 6 - 7 | 585 | 5136664 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
