Assay Details
Gene Symbol: Prdm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027547.2 | NP_081823.2 | 7 - 8 | 992 | 122 | |
GenBank mRNA | AK014501.1 | - | 7 - 8 | 916 | 122 | |
BC138901.1 | - | 7 - 8 | 966 | 8861923 | 122 | |
BC138902.1 | - | 7 - 8 | 966 | 8861924 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
