Assay Details
| Gene Symbol: Suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019879.3 | NP_063932.2 | 2 - 3 | 353 | 76 | |
| GenBank mRNA | AF144101.2 | - | 2 - 3 | 186 | 76 | |
| AK002754.1 | - | 2 - 3 | 188 | 76 | ||
| AK005080.1 | - | 2 - 3 | 202 | 76 | ||
| AK087390.1 | - | 2 - 3 | 191 | 76 | ||
| AK139288.1 | - | 2 - 3 | 181 | 76 | ||
| BC011087.1 | - | 2 - 3 | 174 | 4012310 | 76 | |
| BY061062.1 | - | 2 - 3 | 354 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Suclg1
Genomic Map