Assay Details
| Gene Symbol: Sema4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB021291.1 | - | 1 - 2 | 242 | 62 | |
| AK035993.1 | - | 1 - 2 | 274 | 62 | ||
| AK047067.1 | - | 1 - 2 | 258 | 62 | ||
| AK158292.1 | - | 1 - 2 | 208 | 62 | ||
| BC057048.1 | - | 1 - 2 | 244 | 62 | ||
| BC090625.1 | - | 1 - 2 | 244 | 62 | ||
| BC094567.1 | - | 1 - 2 | 269 | 6834138 | 62 | |
| BC139232.1 | - | 1 - 2 | 216 | 8862254 | 62 | |
| BY237710.1 | - | 1 - 2 | 292 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sema4f
Genomic Map