Assay Details
Gene Symbol: Epha10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256432.1 | NP_001243361.1 | 12 - 13 | 2301 | 105 | |
GenBank mRNA | AK134673.1 | - | 8 - 9 | 929 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
