Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400010801.mm.1 |
NM_001256432.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010801.mm.1 |
NM_001256432.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010801.mm.1 |
TC0400001431.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010804.mm.1 |
NM_001256432.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010804.mm.1 |
NM_001256432.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010804.mm.1 |
TC0400001431.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010802.mm.1 |
NM_001256432.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010802.mm.1 |
NM_001256432.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010802.mm.1 |
TC0400001431.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400010803.mm.1 |
NM_001256432.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010803.mm.1 |
NM_001256432.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400010803.mm.1 |
TC0400001431.mm.1 |
MTA TC |