Assay Details
| Gene Symbol: Cd8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009858.2 | NP_033988.1 | 1 - 2 | 89 | 86 | |
| GenBank mRNA | AK087944.1 | - | 1 - 2 | 66 | 86 | |
| AK088372.1 | - | 1 - 2 | 114 | 86 | ||
| AK088649.1 | - | 1 - 2 | 113 | 86 | ||
| AK154055.1 | - | 1 - 2 | 69 | 86 | ||
| AK162993.1 | - | 1 - 2 | 78 | 86 | ||
| AK169581.1 | - | 1 - 2 | 90 | 86 | ||
| M16799.1 | - | 1 - 2 | 82 | 86 | ||
| M17534.1 | - | 1 - 2 | 89 | 86 | ||
| M19504.1 | - | 1 - 2 | 77 | 86 | ||
| U34882.1 | - | 1 - 2 | 209 | 86 | ||
| X07698.1 | - | 1 - 2 | 93 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd8b1
Genomic Map