Assay Details
| Gene Symbol: Cd8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009858.2 | NP_033988.1 | 5 - 6 | 669 | 81 | |
| GenBank mRNA | AK087944.1 | - | 5 - 6 | 646 | 81 | |
| AK088372.1 | - | 5 - 6 | 694 | 81 | ||
| AK088649.1 | - | 5 - 6 | 693 | 81 | ||
| AK154055.1 | - | 5 - 6 | 649 | 81 | ||
| AK162993.1 | - | 5 - 6 | 658 | 81 | ||
| AK169581.1 | - | 5 - 6 | 670 | 81 | ||
| M16799.1 | - | 5 - 6 | 662 | 81 | ||
| M17534.1 | - | 5 - 6 | 669 | 81 | ||
| M19504.1 | - | 5 - 6 | 657 | 81 | ||
| X07698.1 | - | 5 - 6 | 673 | 81 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154982 | AK162993.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154982 | NM_009858.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154982 | AK088649.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154982 | AK154055.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154982 | AK087944.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47406 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891386 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924090 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 622327 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684536 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183779 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530015 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 711864 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422606 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77405 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114825 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P126691 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154982 | AK162993.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154982 | NM_009858.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154982 | AK088649.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154982 | AK154055.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154982 | AK087944.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P150433 | M17534.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535746 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535748 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535741 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535744 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24535737 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114825 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P126691 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05936012 | - | - |
| Whole Mouse Genome G4122A | A_51_P150433 | M17534.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122499 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100198 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P126691 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114825 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05790900 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05222642 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05032577 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05790899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24535811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535727 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05326929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05200204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05200217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05546878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05202357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05513434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05202355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05754528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05546875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535721 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05326936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05365596 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05326932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05621246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05597296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05365603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05513436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05754531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05790900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05752036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05202358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05032576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05659692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05365598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05546877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05413699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05129082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05413695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05326930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05365600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05679812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05679815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05032577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24535762 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05202354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05621252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05546876 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05621249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05752038 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10538990 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538989 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538985 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538986 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538983 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538987 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538992 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538991 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538984 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426170_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10538979 | - | - |
| Mouse Genome 430 2.0 Array | 1426170_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448569_at | - | - |
| Mouse Genome 430 2.0 Array | 1448569_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1426170_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12526_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12526_PM_at | - | - |
| Mouse Expression Set 430 | 1426170_PM_a_at | - | - |
| Mouse Expression Set 430 | 1426170_a_at | - | - |
| Murine Genome U74v2 Set | 162410_s_at | - | - |
| Murine Genome U74v2 Set | 162410_PM_s_at | - | - |
| Murine Genome U74v2 Set | 94000_PM_at | - | - |
| Murine Genome U74v2 Set | 94000_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426170_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426170_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426170_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426170_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103170408 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1940411 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2570035 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103170408 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002570035 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001940411 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2741169 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2664929 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257694 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2664929 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2741169 | - | - |
| mouse-6 v1.1 (Illumina) | scl26166.1_3-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0012526.1_70-S | - | - |
| mouse-6 v1.1 (Illumina) | scl29902.6.1_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2570035 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1940411 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103170408 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29902.6.1_5-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0012526.1_70-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2570035 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103170408 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1940411 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2664929 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2741169 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257694 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2741169 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2664929 | - | - |

Cd8b1
Genomic Map