Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600019131.mm.1 |
TC0600002473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600019131.mm.1 |
NM_025430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019131.mm.1 |
NM_025430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019129.mm.1 |
TC0600002473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600019129.mm.1 |
NM_025430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019129.mm.1 |
NM_025430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019130.mm.1 |
TC0600002473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600019130.mm.1 |
NM_025430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019130.mm.1 |
NM_025430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019132.mm.1 |
TC0600002473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600019132.mm.1 |
NM_025430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019132.mm.1 |
NM_025430.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019133.mm.1 |
TC0600002473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600019133.mm.1 |
NM_025430.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600019133.mm.1 |
NM_025430.3 |
Refseq |