Assay Details
| Gene Symbol: Thnsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033929.2 | NP_001029101.1 | 1 - 2 | 93 | 104 | |
| GenBank mRNA | CA491740.1 | - | 1 - 2 | 126 | 104 | |
| CX223060.1 | - | 1 - 2 | 93 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001033929.2 transcripts.
Set Membership: |
|

Thnsl2
Genomic Map