Assay Details
| Gene Symbol: Suclg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011507.3 | NP_035637.2 | 5 - 6 | 643 | 70 | |
| GenBank mRNA | AF058956.1 | - | 4 - 5 | 485 | 70 | |
| AK042987.1 | - | 5 - 6 | 598 | 70 | ||
| AK045176.1 | - | 5 - 6 | 596 | 70 | ||
| AK156263.1 | - | 5 - 6 | 598 | 70 | ||
| AK167136.1 | - | 5 - 6 | 598 | 70 | ||
| AK167339.1 | - | 5 - 6 | 882 | 70 | ||
| AK172561.1 | - | 5 - 6 | 598 | 70 | ||
| AK172633.1 | - | 5 - 6 | 598 | 70 | ||
| BC043312.1 | - | 3 - 4 | 338 | 70 | ||
| BC054425.1 | - | 4 - 5 | 396 | 70 | ||
| BC080781.1 | - | 5 - 6 | 598 | 30470296 | 70 | |
| EU234009.1 | - | 5 - 6 | 550 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Suclg2
Genomic Map