Assay Details
Gene Symbol: Slc25a26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026255.5 | NP_080531.2 | 2 - 3 | 308 | 77 | |
GenBank mRNA | BB852792.1 | - | 2 - 3 | 310 | 77 | |
BC019156.1 | - | 3 - 4 | 278 | 5066084 | 77 | |
BC019170.1 | - | 2 - 3 | 309 | 5003084 | 77 | |
BC037142.1 | - | 2 - 3 | 285 | 5026631 | 77 | |
BC037648.1 | - | 1 - 2 | 110 | 77 | ||
BC049899.1 | - | 2 - 3 | 253 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
