Assay Details
| Gene Symbol: Cntn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017383.3 | NP_059079.2 | 14 - 15 | 2097 | 84 | |
| GenBank mRNA | AB032602.1 | - | 13 - 14 | 1783 | 84 | |
| AK052972.1 | - | 15 - 16 | 1990 | 84 | ||
| BC076594.1 | - | 14 - 15 | 2097 | 30649661 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cntn6
Genomic Map