Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600021372.mm.1 |
TC0600002745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021372.mm.1 |
NM_001081146.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021372.mm.1 |
NM_001081146.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021372.mm.1 |
NM_001134459.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021372.mm.1 |
NM_001134459.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021372.mm.1 |
NM_001134460.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021372.mm.1 |
NM_001134461.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021372.mm.1 |
NM_001134461.1 |
Refseq |