Assay Details
| Gene Symbol: Nr2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011630.3 | NP_035760.1 | 13 - 14 | 1919 | 125 | |
| GenBank mRNA | AK157305.1 | - | 13 - 14 | 1893 | 125 | |
| AK220507.1 | - | 6 - 7 | 817 | 125 | ||
| BC103683.1 | - | 11 - 12 | 1538 | 40047943 | 125 | |
| BC103684.1 | - | 11 - 12 | 1538 | 40047942 | 125 | |
| S75970.1 | - | 13 - 14 | 2034 | 125 | ||
| U11688.1 | - | 12 - 13 | 1581 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr2c2
Genomic Map