Assay Details
Gene Symbol: Nr2c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011630.3 | NP_035760.1 | 4 - 5 | 680 | 66 | |
GenBank mRNA | AK157305.1 | - | 4 - 5 | 654 | 66 | |
BC103683.1 | - | 2 - 3 | 299 | 40047943 | 66 | |
BC103684.1 | - | 2 - 3 | 299 | 40047942 | 66 | |
BC103686.1 | - | 2 - 3 | 299 | 40047941 | 66 | |
S75970.1 | - | 4 - 5 | 795 | 66 | ||
U11688.1 | - | 3 - 4 | 342 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
