Assay Details
| Gene Symbol: Nr2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011630.3 | NP_035760.1 | 6 - 7 | 955 | 67 | |
| GenBank mRNA | AK157305.1 | - | 6 - 7 | 929 | 67 | |
| BC103683.1 | - | 4 - 5 | 574 | 40047943 | 67 | |
| BC103684.1 | - | 4 - 5 | 574 | 40047942 | 67 | |
| BC103686.1 | - | 4 - 5 | 574 | 40047941 | 67 | |
| S75970.1 | - | 6 - 7 | 1070 | 67 | ||
| U11688.1 | - | 5 - 6 | 617 | 67 | ||
| U32939.1 | - | 4 - 5 | 588 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr2c2
Genomic Map