Assay Details
| Gene Symbol: Nr2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011630.3 | NP_035760.1 | 8 - 9 | 1202 | 69 | |
| GenBank mRNA | AK220507.1 | - | 1 - 2 | 100 | 69 | |
| BC103683.1 | - | 6 - 7 | 821 | 40047943 | 69 | |
| BC103684.1 | - | 6 - 7 | 821 | 40047942 | 69 | |
| BC103686.1 | - | 6 - 7 | 821 | 40047941 | 69 | |
| S75970.1 | - | 8 - 9 | 1317 | 69 | ||
| U11688.1 | - | 7 - 8 | 864 | 69 | ||
More Information
Set Membership: |
|

Nr2c2
Genomic Map