Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600021710.mm.1 |
TC0600002799.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021710.mm.1 |
NM_001111106.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021710.mm.1 |
NM_011666.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021711.mm.1 |
TC0600002799.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021711.mm.1 |
NM_001111106.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021711.mm.1 |
NM_011666.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021713.mm.1 |
TC0600002799.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021713.mm.1 |
NM_001111106.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021713.mm.1 |
NM_011666.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021712.mm.1 |
TC0600002799.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600021712.mm.1 |
NM_001111106.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600021712.mm.1 |
NM_011666.3 |
Refseq |