Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600008858.mm.1 |
TC0600001136.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600008858.mm.1 |
NM_001113198.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600008858.mm.1 |
NM_001113198.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600008858.mm.1 |
NM_001178049.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600008858.mm.1 |
NM_001178049.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600008858.mm.1 |
NM_008601.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600008858.mm.1 |
NM_008601.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600008857.mm.1 |
TC0600001136.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600008857.mm.1 |
NM_001113198.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600008857.mm.1 |
NM_001113198.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600008857.mm.1 |
NM_001178049.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600008857.mm.1 |
NM_001178049.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600008857.mm.1 |
NM_008601.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600008857.mm.1 |
NM_008601.3 |
Refseq |