Assay Details
| Gene Symbol: Tmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081111.2 | NP_001074580.1 | 4 - 5 | 1671 | 66 | |
| GenBank mRNA | AK144729.1 | - | 4 - 5 | 1672 | 66 | |
| BC150931.1 | - | 4 - 5 | 1682 | 9087842 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmf1
Genomic Map