Assay Details
| Gene Symbol: Tmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081111.2 | NP_001074580.1 | 5 - 6 | 1773 | 82 | |
| GenBank mRNA | AK144729.1 | - | 5 - 6 | 1774 | 82 | |
| BC150931.1 | - | 5 - 6 | 1784 | 9087842 | 82 | |
| CN456538.1 | - | 3 - 4 | 316 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmf1
Genomic Map