Assay Details
Gene Symbol: Klf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023184.3 | NP_075673.1 | 1 - 2 | 102 | 118 | |
GenBank mRNA | AK009739.1 | - | 1 - 2 | 102 | 118 | |
AK138824.1 | - | 1 - 2 | 139 | 118 | ||
AK142618.1 | - | 1 - 2 | 104 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK009739.1, AK142618.1, NM_023184.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
