Assay Details
| Gene Symbol: Ccdc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173775.3 | NP_776136.2 | 15 - 16 | 1786 | 65 | |
| GenBank mRNA | AK048790.1 | - | 7 - 8 | 2381 | 65 | |
| BC047211.1 | - | 13 - 14 | 1468 | 6392183 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc37
Genomic Map