Assay Details
| Gene Symbol: Iqsec1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134383.1 | NP_001127855.1 | 12 - 13 | 3051 | 94 | |
| GenBank mRNA | AK220355.1 | - | 10 - 11 | 1502 | 94 | |
| BC026481.1 | - | 10 - 11 | 1555 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Iqsec1
Genomic Map