Assay Details
Gene Symbol: Sfxn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178639.4 | NP_848754.1 | 7 - 8 | 424 | 68 | |
GenBank mRNA | AF325264.1 | - | 7 - 8 | 442 | 68 | |
AK036285.1 | - | 7 - 8 | 422 | 68 | ||
AK043706.1 | - | 7 - 8 | 423 | 68 | ||
AK082073.1 | - | 7 - 8 | 422 | 68 | ||
BB628882.1 | - | 7 - 8 | 425 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
