Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600022268.mm.1 |
TC0600002903.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022268.mm.1 |
NM_001167730.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022268.mm.1 |
NM_001167730.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022268.mm.1 |
NM_021385.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022268.mm.1 |
NM_021385.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022269.mm.1 |
TC0600002903.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022269.mm.1 |
NM_001167730.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022269.mm.1 |
NM_001167730.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022269.mm.1 |
NM_021385.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022269.mm.1 |
NM_021385.2 |
Refseq |