Assay Details
| Gene Symbol: Rad18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167730.1 | NP_001161202.1 | 4 - 5 | 339 | 90 | |
| NM_021385.2 | NP_067360.2 | 4 - 5 | 339 | 90 | ||
| GenBank mRNA | AF205278.1 | - | 4 - 5 | 263 | 90 | |
| AK012795.1 | - | 4 - 5 | 340 | 90 | ||
| AK030043.1 | - | 4 - 5 | 340 | 90 | ||
| AK048975.1 | - | 4 - 5 | 314 | 90 | ||
| BC011120.1 | - | 4 - 5 | 302 | 4014013 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rad18
Genomic Map