Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 12 - 13 | 1189 | 136 | |
| GenBank mRNA | AK038318.1 | - | 12 - 13 | 1167 | 136 | |
| AK122320.1 | - | 11 - 12 | 1195 | 136 | ||
| AK145716.1 | - | 12 - 13 | 1190 | 136 | ||
| AK161181.1 | - | 12 - 13 | 1139 | 137 | ||
| AK163765.1 | - | 11 - 12 | 1177 | 136 | ||
| AK168388.1 | - | 12 - 13 | 1176 | 136 | ||
| BC056942.1 | - | 12 - 13 | 1116 | 6827177 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam21
Genomic Map