Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 19 - 20 | 1941 | 74 | |
| GenBank mRNA | AK038318.1 | - | 19 - 20 | 1919 | 74 | |
| AK043764.1 | - | 8 - 9 | 786 | 74 | ||
| AK122320.1 | - | 18 - 19 | 1947 | 74 | ||
| AK145716.1 | - | 19 - 20 | 1942 | 74 | ||
| AK161181.1 | - | 19 - 20 | 1891 | 74 | ||
| AK163765.1 | - | 18 - 19 | 1929 | 74 | ||
| AK168388.1 | - | 19 - 20 | 1928 | 74 | ||
| BC032177.1 | - | 17 - 18 | 2168 | 74 | ||
| BC040393.1 | - | 19 - 20 | 1865 | 74 | ||
| BC049979.1 | - | 19 - 20 | 1917 | 74 | ||
| BC056942.1 | - | 19 - 20 | 1868 | 6827177 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam21
Genomic Map