Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 1 - 2 | 92 | 127 | |
| GenBank mRNA | AK011156.1 | - | 1 - 2 | 38 | 127 | |
| AK038318.1 | - | 1 - 2 | 70 | 127 | ||
| AK145716.1 | - | 1 - 2 | 92 | 127 | ||
| AK161181.1 | - | 1 - 2 | 41 | 127 | ||
| AK168388.1 | - | 1 - 2 | 79 | 127 | ||
| BC049979.1 | - | 1 - 2 | 68 | 127 | ||
| BY768668.1 | - | 1 - 2 | 93 | 127 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011156.1, AK038318.1, BC049979.1, NM_026585.3 transcripts.
Set Membership: |
|

Fam21
Genomic Map