Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 25 - 26 | 2771 | 131 | |
| GenBank mRNA | AK043764.1 | - | 14 - 15 | 1616 | 131 | |
| AK145716.1 | - | 25 - 26 | 2772 | 131 | ||
| AK161181.1 | - | 25 - 26 | 2721 | 131 | ||
| AK168388.1 | - | 25 - 26 | 2760 | 132 | ||
| BC032177.1 | - | 23 - 24 | 2997 | 132 | ||
| BC056942.1 | - | 25 - 26 | 2698 | 6827177 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam21
Genomic Map