Assay Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026585.3 | NP_080861.2 | 4 - 5 | 444 | 77 | |
| GenBank mRNA | AK011156.1 | - | 4 - 5 | 390 | 77 | |
| AK038318.1 | - | 4 - 5 | 422 | 77 | ||
| AK122320.1 | - | 3 - 4 | 450 | 77 | ||
| AK145716.1 | - | 4 - 5 | 444 | 77 | ||
| AK161181.1 | - | 4 - 5 | 393 | 77 | ||
| AK163765.1 | - | 3 - 4 | 432 | 77 | ||
| AK168388.1 | - | 4 - 5 | 431 | 77 | ||
| BC032177.1 | - | 2 - 3 | 671 | 77 | ||
| BC040393.1 | - | 4 - 5 | 369 | 77 | ||
| BC049979.1 | - | 4 - 5 | 420 | 77 | ||
| BC056942.1 | - | 4 - 5 | 371 | 6827177 | 77 | |
More Information
Set Membership: |
|

Fam21
Genomic Map