Assay Details
| Gene Symbol: Zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290339.1 | NP_001277268.1 | 1 - 2 | 49 | 108 | |
| GenBank mRNA | AK012639.1 | - | 1 - 2 | 50 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290339.1 transcripts.
Set Membership: |
|

Zfand4
Genomic Map