Assay Details
| Gene Symbol: Zfand4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290338.1 | NP_001277267.1 | 6 - 7 | 1969 | 68 | |
| NM_001290339.1 | NP_001277268.1 | 4 - 5 | 1669 | 68 | ||
| GenBank mRNA | AK006693.1 | - | 1 - 2 | 576 | 68 | |
| AK012639.1 | - | 4 - 5 | 1668 | 68 | ||
| AK089032.1 | - | 6 - 7 | 1966 | 68 | ||
| CX223310.1 | - | 1 - 2 | 101 | 68 | ||
More Information
Set Membership: |
|

Zfand4
Genomic Map