Assay Details
| Gene Symbol: Usp39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138592.4 | NP_613058.1 | 9 - 10 | 1304 | 78 | |
| GenBank mRNA | AK010443.1 | - | 9 - 10 | 1314 | 78 | |
| AK018595.1 | - | 9 - 10 | 1305 | 78 | ||
| AK033105.1 | - | 9 - 10 | 1301 | 78 | ||
| AK045062.1 | - | 9 - 10 | 1301 | 78 | ||
| AK088354.1 | - | 9 - 10 | 1316 | 78 | ||
| AK165625.1 | - | 9 - 10 | 1285 | 78 | ||
| AK167668.1 | - | 9 - 10 | 1292 | 78 | ||
| BC026983.1 | - | 9 - 10 | 1279 | 5365862 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp39
Genomic Map