Assay Details
Gene Symbol: Retsat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026159.4 | NP_080435.3 | 1 - 2 | 163 | 61 | |
GenBank mRNA | AK144115.1 | - | 319 | 61 | ||
AY704159.1 | - | 1 - 2 | 163 | 61 | ||
BC117750.1 | - | 1 - 2 | 158 | 61 | ||
BC117751.1 | - | 1 - 2 | 163 | 40090531 | 61 | |
EF620363.1 | - | 189 | 61 | |||
EF620364.1 | - | 189 | 61 | |||
EF620365.1 | - | 1 - 2 | 189 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
