Assay Details
| Gene Symbol: Retsat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026159.4 | NP_080435.3 | 8 - 9 | 1362 | 69 | |
| GenBank mRNA | AK002851.1 | - | 5 - 6 | 714 | 69 | |
| AK009103.1 | - | 4 - 5 | 417 | 69 | ||
| AK144115.1 | - | 69 | ||||
| AY704159.1 | - | 8 - 9 | 1362 | 69 | ||
| BC011203.1 | - | 4 - 5 | 544 | 69 | ||
| BC117750.1 | - | 8 - 9 | 1357 | 69 | ||
| BC117751.1 | - | 8 - 9 | 1362 | 40090531 | 69 | |
| EF620363.1 | - | 1388 | 69 | |||
| EF620364.1 | - | 1388 | 69 | |||
| EF620365.1 | - | 8 - 9 | 1388 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Retsat
Genomic Map