Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600024842.mm.1 |
TC0600003185.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024842.mm.1 |
NM_175414.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024842.mm.1 |
NM_175414.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024851.mm.1 |
TC0600003185.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024851.mm.1 |
NM_175414.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024851.mm.1 |
NM_175414.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024841.mm.1 |
TC0600003185.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024841.mm.1 |
NM_175414.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024841.mm.1 |
NM_175414.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024846.mm.1 |
TC0600003185.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600024846.mm.1 |
NM_175414.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600024846.mm.1 |
NM_175414.4 |
Refseq |