Assay Details
| Gene Symbol: Sumf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145937.3 | NP_666049.2 | 5 - 6 | 744 | 76 | |
| GenBank mRNA | AK151874.1 | - | 5 - 6 | 745 | 76 | |
| AK159192.1 | - | 5 - 6 | 733 | 76 | ||
| AK160917.1 | - | 5 - 6 | 726 | 76 | ||
| AK161203.1 | - | 5 - 6 | 737 | 76 | ||
| AK216554.1 | - | 1 - 2 | 76 | |||
| BC026981.1 | - | 5 - 6 | 717 | 5365580 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sumf1
Genomic Map