Assay Details
| Gene Symbol: Pold2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008894.2 | NP_032920.2 | 10 - 11 | 1368 | 84 | |
| GenBank mRNA | AK013901.1 | - | 10 - 11 | 1333 | 84 | |
| BC003342.1 | - | 10 - 11 | 1334 | 3257610 | 84 | |
| BM215242.2 | - | 1 - 2 | 84 | |||
| Z72486.1 | - | 10 - 11 | 1329 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565429 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565426 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342707 | CB205181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342707 | Z72486.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342707 | AK013901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342707 | BC003342.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342707 | BM215242.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342707 | CB205181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342707 | Z72486.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342707 | AK013901.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342707 | BC003342.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P342707 | BM215242.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 237475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377487 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54211 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22862 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148828 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342707 | CB205181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342707 | Z72486.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342707 | AK013901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342707 | BC003342.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342707 | BM215242.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342707 | CB205181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342707 | Z72486.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342707 | AK013901.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342707 | BC003342.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P342707 | BM215242.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342707 | CB205181.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342707 | Z72486.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342707 | AK013901.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342707 | BC003342.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342707 | BM215242.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342707 | CB205181.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342707 | Z72486.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342707 | AK013901.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342707 | BC003342.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P342707 | BM215242.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565435 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565431 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565443 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565432 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565422 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565444 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565433 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565425 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565420 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565428 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565429 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27565430 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P148828 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07244342 | - | - |
| Whole Mouse Genome G4122A | A_51_P342707 | CB205181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342707 | Z72486.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342707 | AK013901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342707 | BC003342.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342707 | BM215242.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342707 | CB205181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342707 | Z72486.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342707 | AK013901.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342707 | BC003342.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P342707 | BM215242.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109343 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124850 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P148828 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09032003 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09032002 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09032000 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09460453 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565428 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565431 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565427 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565426 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565430 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27565429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27565430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09419949 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565407 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09032003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09058246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09419946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565389 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565401 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09121429 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565409 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565440 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09419941 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09317546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565420 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565447 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09228570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09032002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09398370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565428 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565422 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565411 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09460453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09032000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27565452 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10384033 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384036 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384038 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384039 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384035 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384042 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384034 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384041 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384040 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384043 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10384037 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448277_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10384032 | - | - |
| Mouse Genome 430 2.0 Array | 1448277_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448277_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18972_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18972_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100020937.mm.1 | NM_008894.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100020937.mm.1 | NM_008894.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100020937.mm.1 | TC1100002155.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100020938.mm.1 | NM_008894.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100020938.mm.1 | NM_008894.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100020938.mm.1 | TC1100002155.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1448277_PM_at | - | - |
| Mouse Expression Set 430 | 1448277_at | - | - |
| Murine Genome U74v2 Set | 98155_PM_r_at | - | - |
| Murine Genome U74v2 Set | 98155_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448277_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448277_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448277_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448277_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 346803 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6400148 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006400148 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2765789 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2765789 | - | - |
| mouse-6 v1.1 (Illumina) | scl40552.11.1_156-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6400148 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40552.11.1_156-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6400148 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2817892 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2817892 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40552.11.1_156-S | - | - |

Pold2
Genomic Map