Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100020944.mm.1 |
NM_008894.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020944.mm.1 |
NM_008894.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020944.mm.1 |
TC1100002155.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020942.mm.1 |
NM_008894.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020942.mm.1 |
NM_008894.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020942.mm.1 |
TC1100002155.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020943.mm.1 |
NM_008894.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020943.mm.1 |
NM_008894.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020943.mm.1 |
TC1100002155.mm.1 |
MTA TC |