Assay Details
Gene Symbol: Gck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287386.1 | NP_001274315.1 | 5 - 6 | 701 | 63 | |
NM_010292.5 | NP_034422.2 | 5 - 6 | 1059 | 63 | ||
GenBank mRNA | AK142698.1 | - | 5 - 6 | 700 | 63 | |
BC011139.1 | - | 5 - 6 | 690 | 4195943 | 63 | |
L38990.1 | - | 5 - 6 | 577 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
