Assay Details
| Gene Symbol: Bid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007544.3 | NP_031570.2 | 3 - 4 | 361 | 71 | |
| GenBank mRNA | AK045731.1 | - | 3 - 4 | 376 | 71 | |
| AK051076.1 | - | 377 | 71 | |||
| AK052356.1 | - | 3 - 4 | 366 | 71 | ||
| AK077657.1 | - | 3 - 4 | 344 | 71 | ||
| AK161235.1 | - | 3 - 4 | 383 | 71 | ||
| BC002031.1 | - | 3 - 4 | 361 | 3486537 | 71 | |
| U75506.1 | - | 2 - 3 | 238 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bid
Genomic Map